Learn More
ACROBiosystems Biotinylated Human LRP-6, His,Avitag (LR6-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Pro 630 (Accession # O75581-1 ).
Supplier:  ACROBiosystems LR6H82E7200UG
Encompass_Preferred
LRP6 is a transmembrane low-density lipoprotein receptor that shares a similar structure with LRP5. Low-density lipoprotein receptor-related protein 6 is a key component of the LRP5/LRP6/Frizzled co-receptor group that is involved in canonical Wnt pathway. The Wnt-induced Fzd/LRP6 coreceptor complex recruits DVL1 polymers to the plasma membrane which, in turn, recruits the AXIN1/GSK3B-complex to the cell surface promoting the formation of signalsomes and inhibiting AXIN1/GSK3-mediated phosphorylation and destruction of beta-catenin.
Catalog No. 50-223-0718