Learn More
Sino Biological Anti-LRP10 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13228T1620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LRP10 (rh LRP10; Catalog#13228-H08H; Q7Z4F1-1; Met1-Lys440). LRP10 specific IgG was purified by Human LRP10 affinity chromatography.
Catalog No. 50-241-1094