Learn More
Sino Biological Recombinant Human LRP-6 Protein (hFc Tag)
Catalog No. 502993468
Encompass_Preferred
A DNA sequence encoding the Human LRP6 (O75581) (Ala20-Pro630) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-299-3468 Supplier Sino Biological Supplier No. 11968H02H1100
A DNA sequence encoding the Human LRP6 (O75581) (Ala20-Pro630) was expressed with the Fc region of human IgG1 at the C-terminus.