Learn More
Sino Biological Recombinant Human LRP-6 Protein (His Tag)
Catalog No. 502993562
Encompass_Preferred
A DNA sequence encoding the Human LRP6 (O75581-1) (Ala19-Pro630) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3562 Supplier Sino Biological Supplier No. 17052H08H1100
A DNA sequence encoding the Human LRP6 (O75581-1) (Ala19-Pro630) was expressed with a polyhistidine tag at the C-terminus.