Learn More
Sino Biological Recombinant Human LRP-6 Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 11968H49HB20
Encompass_Preferred
A DNA sequence encoding the Human LRP6 (O75581) (Met1-Pro630) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4480