Learn More
Sino Biological Human LRPAP1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11100MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LRPAP1 / A2MRAP (rh LRPAP1; Catalog#11100-H08H; Tyr 35-Leu 357; NP_002328.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1607