Learn More
Sino Biological Anti-LRPAP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11100RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LRPAP1 / A2MRAP (rh LRPAP1; Catalog#11100-H08H; NP_002328.1; Tyr 35-Leu 357). LRPAP1 specific IgG was purified by human LRPAP1 affinity chromatography.
Catalog No. 50-241-0585