Learn More
ACROBiosystems Human LRRC15 / LIB Protein, His Tag
Catalog No. 502054802
Encompass_Preferred
Recombinant Protein;100UG;Human LRRC15, His Tag (LR5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Tyr 22 - Gly 538 (Accession # Q8TF66-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.7 kDa.
Catalog No. 50-205-4802 Supplier ACROBiosystems Supplier No. LR5H52H3100UG
Recombinant Protein;100UG;Human LRRC15, His Tag (LR5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Tyr 22 - Gly 538 (Accession # Q8TF66-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.7 kDa.