Learn More
ACROBiosystems Human LRRC15 / LIB Protein, His Tag
Supplier:  ACROBiosystems LR5H52H31MG
Encompass_Preferred
Recombinant Protein;1MG;Human LRRC15, His Tag (LR5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Tyr 22 - Gly 538 (Accession # Q8TF66-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.7 kDa.
Catalog No. 50-205-4801