Learn More
Sino Biological Recombinant Human LRRC15 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 15786H08H100
Encompass_Preferred
A DNA sequence encoding the human LRRC15 (NP_570843.2) (Met1-Gly538) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6622