Learn More
Sino Biological Recombinant Human LRRC32 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 15508H08HB20
Encompass_Preferred
A DNA sequence encoding the human LRRC32 (NP_005503.1) (Met1-Asn627) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4097