Learn More
Sino Biological Anti-NGL-1/LRRC4C Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200556T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human NGL-1/LRRC4C fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3963