Learn More
Sino Biological Anti-LSM14A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102713T40100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human LSM14A, and purified by antigen affinity chromatography.
Catalog No. 50-241-2378