Learn More
Sino Biological Anti-Lymphotoxin alpha/TNF beta Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10270MM0220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TNF-beta / TNFSF1 / Lymphotoxin alpha (rh TNF-beta / TNFSF1 / Lymphotoxin alpha ; Catalog#10270-HNAE; P01374; Leu 35-Leu 205). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1499