Learn More
Raybiotech Inc Human LTA4H ELISA Kit
Supplier:  Raybiotech Inc ELHLTA4H1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 2.1 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects Human LTA4H.
Catalog No. 50-194-9512