Learn More
Abcam Human LTBP2 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB3139881X96TESTS
Encompass_Preferred
Human LTBP2 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human LTBP2 in Human in Serum, Plasma, Cell culture supernatant samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2814179