Learn More
BIOHIPPO Human LTBP2 Microsample
SDP
Catalog No. 5000315518
Encompass
Human LTBP2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LTBP2 in serum plasma urine tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 127 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human LTBP2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human LTBP2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human LTBP2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15518 Supplier BIOHIPPO Supplier No. BHE15212111
Human LTBP2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LTBP2 in serum plasma urine tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 127 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human LTBP2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human LTBP2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human LTBP2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color