Learn More
Sino Biological Anti-LTC4S Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11622RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LTC4S (rh LTC4S; Catalog#11622-H08B; NP_665874.1; Met1-Ala150). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4661