Learn More
Raybiotech Inc Human Lactoferrin ELISA Kit
Supplier:  Raybiotech Inc ELHLTF1
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 1.96 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human Adipsin, Angiogenin, B2M, bIG-H3, CD14, CRP, DPPIV, Endostatin, E-Selectin, FAP, Ferritin, Fetuin A, gp130, HGFR, ICAM-2, IGFBP-2, IGFBP-3, IGFBP-6, Lipocalin-2, L-Selectin, LYVE-1, MCSF R, MMP-9, NCAM-1, Nidogen-1, PAI-1, PF4, RBP4, SAA, TSP-1.
Catalog No. 50-194-9514