Learn More
Sino Biological Recombinant Anti-Lactoferrin/LTF Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 11096R006H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Lactotransferrin / LTF (rh Lactotransferrin / LTF; Catalog#11096-H08H; P02788; Met 1-Lys 710), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-8627