Learn More
BIOHIPPO Human LTF Microsample
SDP
Catalog No. 5000315769
Encompass
Human LTF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LTF in serum plasma tissue homogenates cell lysates breast milk cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 ng/mL with a sensitivity of 21 3 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human LTF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human LTF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human LTF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15769 Supplier BIOHIPPO Supplier No. BHE15212679
Human LTF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LTF in serum plasma tissue homogenates cell lysates breast milk cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 ng/mL with a sensitivity of 21 3 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human LTF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human LTF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human LTF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color