Learn More
Sino Biological Anti-Lumican Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11640RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LUM (rh LUM; Catalog#11640-H08H; NP_002336.1; Met 1-Asn 338). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4653