Learn More
Abcam Human Lumican ELISA Kit, 1 x 96 Tests
Catalog No. 502698637
Encompass_Preferred
Human Lumican ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Lumican with a sensitivity of 20.177 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-8637 Supplier Abcam Supplier No. AB3150541X96TESTS
Human Lumican ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Lumican with a sensitivity of 20.177 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement