Learn More
Sino Biological Human LXN Protein 50ug (Sino Biological)
Supplier:  Sino Biological 11044H07EP100
Encompass_Preferred
A DNA sequence encoding the human Latexin (NP_064554.3) (Glu 2-Glu222) was expressed, fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-4135