Learn More
Invitrogen™ Human LY6G5B (aa 98-192) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (63%), Rat (63%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61705. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q8NDX9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Neutralization, Control |
| Formulation | PBS, 1M urea with no preservative; pH 7.4 |
| Gene ID (Entrez) | 58496 |
| Name | Human LY6G5B (aa 98-192) Control Fragment |
| pH Range | 7.4 |
| Purification Method | Purified |
| Quantity | 100 μL |
| Storage Requirements | -20°C, Avoid Freeze/Thaw Cycles |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.