Learn More
Sino Biological Recombinant Human LY6G6D Protein (hFc Tag)
Supplier:  Sino Biological 24266H01H100
Encompass_Preferred
A DNA sequence encoding the Human LY6G6D (O95868) (Asn20-Asn103) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-255-3857