Learn More
ACROBiosystems Human LY75 / CD205 Protein, His Tag
Catalog No. 502054859
Encompass_Preferred
Recombinant Protein;1MG;Human LY75, His Tag (LY5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Asp 1666 (Accession # O60449-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 190.7 kDa.
Catalog No. 50-205-4859 Supplier ACROBiosystems Supplier No. LY5H52H31MG
Recombinant Protein;1MG;Human LY75, His Tag (LY5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Asp 1666 (Accession # O60449-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 190.7 kDa.