Learn More
Sino Biological Recombinant Human LY75/CD205 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 16490H08H1
Encompass_Preferred
A DNA sequence encoding the human LY75 (NP_002340.2) (Ala32-Gly1664) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4026