Learn More
Sciencell Research Laboratories Human Lymphatic Endothelial Cells, Passage one, >5 x 10⁵ cells per 1 ml vial, Characterization markers: vWF/Factor VIII, CD31, Liquid nitrogen storage, Dry ice shipping
Supplier:  Sciencell Research Laboratories 2500
Encompass
Human Lymphatic Endothelial Cells (HLEC) are primary cells isolated from human lymph nodes that play a critical role in the study of the lymphatic and immune systems. These cells are vital for maintaining tissue homeostasis and are crucial for researching immune responses, lymphocyte behavior, chronic inflammation, and cancer research. Supplied cryopreserved at passage one, HLEC exhibit high viability, making them ideal for a variety of in vitro applications.
  • Physiologically relevant model for investigating lymphatic endothelium function
  • High quality and purity with rigorous quality control
  • Optimized for excellent proliferative capacity and consistent performance
  • Negative for common contaminants such as HIV-1, HBV, and HCV
  • More than 5 x 10⁵ cells per 1 ml vial
  • Guaranteed expansion of 10 population doublings
  • Storage in liquid nitrogen with shipping via dry ice
  • Recommended seeding density of 5,000-7,000 cells/cm²
Catalog No. NC2391449