Learn More
Invivogen Raji-hPD-1 Cells, Human Lymphoblast-Like Cells, Target Cells for ADCC Assays, 3-7 x 10^6 cells
SDP
Catalog No. NC1775561
Encompass
Raji-hPD-1 cells are engineered human lymphoblast-like cells designed for in vitro assays, especially antibody-dependent cellular cytotoxicity (ADCC) studies. They feature stable overexpression of human PD-1, verified by flow cytometry. These cells serve as target cells for ADCC and CAR-T cell toxicity assays, supporting functional immune checkpoint research. Their guaranteed mycoplasma-free status and high passage stability make them reliable for experiments.
Catalog No. NC1775561 Supplier Invivogen Supplier No. RAJIHPD1
Raji-hPD-1 cells are engineered human lymphoblast-like cells designed for in vitro assays, especially antibody-dependent cellular cytotoxicity (ADCC) studies. They feature stable overexpression of human PD-1, verified by flow cytometry. These cells serve as target cells for ADCC and CAR-T cell toxicity assays, supporting functional immune checkpoint research. Their guaranteed mycoplasma-free status and high passage stability make them reliable for experiments.