Learn More
Bioworld HUMAN LYMPHOTACTIN (XCL1)
Supplier:  Bioworld 220604111
Encompass_Preferred
HUMAN LYMPHOTACTIN (XCL1), (RECOMBINANT): Recombinant Human Lymphotactin (XCL1) Source:E.Coli Description : Recombinant Human Lymphotactin produced in E.Coli is a single,non-glycosylated, polypeptide chain containing 92 amino acids a...
Catalog No. 50-253-507