Learn More
Sino Biological Human Lymphotoxin alpha/TNF beta Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786336
Encompass_Preferred
The Human TNF-beta / TNFSF1 / Lymphotoxin alpha ELISA Pair Set is for the quantitative determination of Human TNF-beta / TNFSF1 / Lymphotoxin alpha.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human TNF-beta / TNFSF1 / Lymphotoxin alpha coated on a 96-well plate. Standards and samples are added to the wells, and any Human TNF-beta / TNFSF1 / Lymphotoxin alpha present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human TNF-beta / TNFSF1 / Lymphotoxin alpha monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human TNF-beta / TNFSF1 / Lymphotoxin alpha present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at
Catalog No. 50-007-86336 Supplier Sino Biological Supplier No. SEK1027015
The Human TNF-beta / TNFSF1 / Lymphotoxin alpha ELISA Pair Set is for the quantitative determination of Human TNF-beta / TNFSF1 / Lymphotoxin alpha.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human TNF-beta / TNFSF1 / Lymphotoxin alpha coated on a 96-well plate. Standards and samples are added to the wells, and any Human TNF-beta / TNFSF1 / Lymphotoxin alpha present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human TNF-beta / TNFSF1 / Lymphotoxin alpha monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human TNF-beta / TNFSF1 / Lymphotoxin alpha present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at