Learn More
Biomatik Corporation Human Lymphotoxin Beta (LTb) CLIA Kit
Supplier:  Biomatik Corporation EKU09669
Encompass_Preferred
Human Lymphotoxin Beta (LTb) CLIA Kit for detecting LTb in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.49pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q06643. Gene ID: 4050. Target Synonyms: LT-B; p33; TNFC; TNF-C; TNFSF3; Tumor necrosis factor ligand superfamily member 3; Tumor Necrosis Factor C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6870