Learn More
ACROBiosystems ACROBiosystems Human LYPD3 / C4.4A Protein, His Tag
Supplier:  ACROBiosystems LY3H52H51MG
Encompass_Preferred
Recombinant Protein;1MG;Human LYPD3, His Tag (LY3-H52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 31 - His 286 (Accession # AAH39167).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.7 kDa. The protein migrates as 45-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6713