Learn More
Biomatik Corporation Human Lysine Specific Demethylase 4A (KDM4A) ELISA Kit
Supplier:  Biomatik Corporation EKU05710
Encompass_Preferred
Human Lysine Specific Demethylase 4A (KDM4A) ELISA Kit for detecting KDM4A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.126ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75164. Gene ID: 9682. Target Synonyms: JMJD2; JMJD2A; JHDM3A; JmjC domain-containing histone demethylation protein 3A; Jumonji domain-containing protein 2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6539