Learn More
Cusabio Technology LLC HUMAN lysyl oxidase ELISA KIT
Catalog No. 5000052057
Encompass_Preferred
This kit detects LOX (Protein-lysine 6-oxidase) in human serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a dynamic range of 0 625 ng/mL to 40 ng/mL with a sensitivity of 0 156 ng/mL enabling precise measurement of low-abundance targets The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours making it suitable for high-throughput applications Detection is performed at 450 nm ensuring reliable optical readouts Designed for research and diagnostic use this kit provides robust performance for analyzing LOX levels in diverse biological matrices while maintaining consistency across experimental conditions
Catalog No. 50-000-52057 Supplier Cusabio Technology LLC Supplier No. CSBE16376H
This kit detects LOX (Protein-lysine 6-oxidase) in human serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a dynamic range of 0 625 ng/mL to 40 ng/mL with a sensitivity of 0 156 ng/mL enabling precise measurement of low-abundance targets The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours making it suitable for high-throughput applications Detection is performed at 450 nm ensuring reliable optical readouts Designed for research and diagnostic use this kit provides robust performance for analyzing LOX levels in diverse biological matrices while maintaining consistency across experimental conditions