Learn More
American Research Products Inc Human Lysyl oXidase homolog 1
Catalog No. 5000327447
Encompass
Human Lysyl oXidase homolog 1 (LOXL1) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-27447 Supplier American Research Products Inc Supplier No. CSBEL013040HU
Human Lysyl oXidase homolog 1 (LOXL1) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates Method Sandwich