Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Macrophage Colony Stimulating Factor 1, His Tag, Lyophilized, 50 μg

Catalog No. 502106717
Encompass_Preferred

Macrophage colony-stimulating factor 1 (M-CSF/CSF-1) is a secreted cytokine and hematopoietic growth factor that regulates the differentiation, proliferation, and survival of monocytes, macrophages, and bone marrow progenitor cells. This human M-CSF protein is expressed from HEK293 cells and contains an N-terminal polyhistidine tag. It is supplied as a lyophilized powder.

  • Expressed in human HEK293 cells
  • Contains N-terminal polyhistidine tag
  • Purity >95% as determined by SDS-PAGE
  • Endotoxin <1.0 EU per μg by LAL method
  • Calculated molecular weight of 26.0 kDa (migrates as 36-45 kDa due to glycosylation)
  • Formulated in PBS, pH 7.4 with trehalose

Catalog No. 50-210-6717 Supplier ACROBiosystems Supplier No. MCFH524750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Macrophage colony-stimulating factor 1 (M-CSF/CSF-1) is a secreted cytokine and hematopoietic growth factor that regulates the differentiation, proliferation, and survival of monocytes, macrophages, and bone marrow progenitor cells. This human M-CSF protein is expressed from HEK293 cells and contains an N-terminal polyhistidine tag. It is supplied as a lyophilized powder.

  • Expressed in human HEK293 cells
  • Contains N-terminal polyhistidine tag
  • Purity >95% as determined by SDS-PAGE
  • Endotoxin <1.0 EU per μg by LAL method
  • Calculated molecular weight of 26.0 kDa (migrates as 36-45 kDa due to glycosylation)
  • Formulated in PBS, pH 7.4 with trehalose

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.