Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human M-CSF Quantikine ELISA Kit

Catalog No. SMC00B
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
SMC00B 6 Plates 6 x 96-well Plates
DMC00B 1 Plate 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SMC00B Supplier R&D Systems™ Supplier No. SMC00B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human M-CSF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human M-CSF Quantikine ELISA Kit quantifies human M-CSF.

Specifications

Assay Range 78.10 to 5000 pg/mL
Assay Sensitivity 47.3 pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Specificity Natural and Recombinant Human M-CSF
Sufficient For 6 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 1435
Quantity 6 Plates
Sample Volume Cell Culture Supernates (100 μL), Serum (100 μL), EDTA Plasma (100 μL), Heparin Plasma (100 μL), Saliva (100 μL), Urine (100 μL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.