Learn More
ACROBiosystems Human M-CSF R / CSF1R / CD115 Protein, Mouse IgG2a Fc Tag (MALS verified)
Supplier:  ACROBiosystems CSRH52551MG
Encompass_Preferred
Recombinant Protein;1MG;Human M-CSF R, Mouse IgG2a Fc Tag (MALS verified) (CSR-H5255) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 (Accession # P07333-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 81.4 kDa. The protein migrates as 105-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human M-CSF R, Mouse IgG2a Fc Tag (MALS verified) (Cat. No. CSR-H5255) at 5 μg/mL (100 μL/well) can bind Biotinylated Human M-CSF, His,Avitag (Cat. No. MCF-H82E6) with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-205-5110