Learn More
Promocell GmbH PromoCell hMDM-MCSF(-)-c 1,5 million Human M2 Macrophages (M-CSF), Monocyte-derived, single donor, cryopreserved
Catalog No. NC1425697
Encompass
PromoCell's crypreserved human monocyte-derived macrophages (hMDM) are convenient and easy-to-handle. The thawed cells plate into all tissue culture vessel formats and can be maintained as adherent, biologically functional cultures for several weeks. Optionally, user-customizable activation of the cells can be performed.Cryopreserved human macrophages are produced from monocytes in PromoCell's well-proven M2 Macrophage Generation Media XF and are available as non-activated, fully qualified M2- (M-CSF-) polarized cells. After 9-10 days of differentiation, the M2 macrophages are cryopreserved using PromoCells's proprietary, defined and animal-free freezing medium, Cryo-SFM. Each cryo vial contains more than 1.5 million or 5 million viable cells after thawing, respectively.Our M2 Macrophages are now also available from HLA-typed donors.
Catalog No. NC1425697 Supplier Promocell GmbH Supplier No. C12915
PromoCell's crypreserved human monocyte-derived macrophages (hMDM) are convenient and easy-to-handle. The thawed cells plate into all tissue culture vessel formats and can be maintained as adherent, biologically functional cultures for several weeks. Optionally, user-customizable activation of the cells can be performed.Cryopreserved human macrophages are produced from monocytes in PromoCell's well-proven M2 Macrophage Generation Media XF and are available as non-activated, fully qualified M2- (M-CSF-) polarized cells. After 9-10 days of differentiation, the M2 macrophages are cryopreserved using PromoCells's proprietary, defined and animal-free freezing medium, Cryo-SFM. Each cryo vial contains more than 1.5 million or 5 million viable cells after thawing, respectively.Our M2 Macrophages are now also available from HLA-typed donors.