Learn More
Biomatik Corporation Human Macrophage Derived Chemokine (MDC) CLIA Kit
Supplier:  Biomatik Corporation EKU08821
Encompass_Preferred
Human Macrophage Derived Chemokine (MDC) CLIA Kit for detecting MDC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 11.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O00626. Gene ID: 6367. Target Synonyms: CCL22; ABCD1; DC/B-CK; SCYA22; STCP1; Chemokine C-C-Motif Ligand 22; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 22; Stimulated T-Cell Chemotactic Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6819