Learn More
Biomatik Corporation Human Macrophage Inflammatory Protein 1 Alpha (MIP1a) CLIA Kit
Supplier:  Biomatik Corporation EKU08823
Encompass_Preferred
Human Macrophage Inflammatory Protein 1 Alpha (MIP1a) CLIA Kit for detecting MIP1a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.95pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P10147. Gene ID: 6348. Target Synonyms: CCL3; MIP1-A; SCYA3; Chemokine C-C-Motif Ligand 3; Small Inducible Cytokine A3; Homologous To Mouse Mip-1a; Tonsillar Lymphocyte LD78 Alpha Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6788