Learn More
Biomatik Corporation Human Macrophage Scavenger Receptor 1 (MSR1) CLIA Kit
Supplier:  Biomatik Corporation EKU09775
Encompass_Preferred
Human Macrophage Scavenger Receptor 1 (MSR1) CLIA Kit for detecting MSR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 11.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P21757. Gene ID: 4481. Target Synonyms: CD204; MSR1; SCARA1; SR-A; PhSR1; PhSR2; Macrophage acetylated LDL receptor I and II; Scavenger receptor class A member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8438