Learn More
Biomatik Corporation Human Macrophage-Derived Chemokine (Mdc/Ccl22) Elisa Kit
Supplier:  Biomatik Corporation EKC31278
Encompass_Preferred
Human Macrophage-Derived Chemokine (Mdc/Ccl22) Elisa Kit for detecting Chemokine (C-C Motif) Ligand 22 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 10.324pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O00626; CCL22. Target Synonyms: A 152E5.1; ABCD 1; ABCD1; C C motif chemokine 22; CC chemokine STCP 1; CC chemokine STCP-1; ccl 22; Ccl22; CCL22_HUMAN; Chemokine (C C motif) ligand 22; DC/B CK; DCBCK; Macrophage-derived chemokine; MDC; MDC (1-69); MDC (7-69); MGC34554; SCYA22; Small inducible cytokine subfamily A (Cys Cys) member 22; Small inducible cytokine subfamily A; member 22; Small-inducible cytokine A22; STCP 1; STCP1; Stimulated T cell chemotactic protein 1; Stimulated T-cell chemotactic protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6610