Learn More
ACROBiosystems Biotinylated Human MAdCAM-1 Protein, Fc,Avitag™ (MALS verified)
Catalog No. 502331727
Encompass_Preferred
Biotinylated Human MAdCAM-1, Fc Tag (MAM-H8251) is expressed from human 293 cells (HEK293). It contains AA Gln 19- Gln 333 (Accession # Q4PKD0-1).
Catalog No. 50-233-1727 Supplier ACROBiosystems Supplier No. MAMH825125UG
Biotinylated Human MAdCAM-1, Fc Tag (MAM-H8251) is expressed from human 293 cells (HEK293). It contains AA Gln 19- Gln 333 (Accession # Q4PKD0-1).