Learn More
BIOHIPPO Human MAG Microsample
SDP
Catalog No. 5000315845
Encompass
Human MAG Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MAG in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 78-50 ng/mL with a sensitivity of 0 333 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MAG Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MAG Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MAG biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15845 Supplier BIOHIPPO Supplier No. BHE15212033
Human MAG Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MAG in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 78-50 ng/mL with a sensitivity of 0 333 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MAG Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MAG Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MAG biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color