Learn More
Raybiotech Inc Human MAGP-2/MFAP5 ELISA Kit
Supplier:  Raybiotech Inc ELHMAGP21
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 40 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human MAGP-2.
Catalog No. 50-194-9519