Learn More
Biomatik Corporation Human Malate Dehydrogenase 1 (MDH1) ELISA Kit
Supplier:  Biomatik Corporation EKU05779
Encompass_Preferred
Human Malate Dehydrogenase 1 (MDH1) ELISA Kit for detecting MDH1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P40925. Gene ID: 4190. Target Synonyms: MDHA; Malate Dehydrogenase 1,NAD (Soluble) ; Cytosolic malate dehydrogenase; Diiodophenylpyruvate reductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8886