Learn More
AURORA BIOLABS LLC Human Malic enzyme 2 (ME2), 100 μg
Catalog No. NC3787982
Encompass
Mitochondrial NAD-dependent malic enzyme (ME2) is a homotetrameric protein that catalyzes the oxidative decarboxylation of malate to pyruvate. It had previously been weakly linked to a syndrome known as Friedreich ataxia that has since been shown to be the result of mutation in a completely different gene. Certain single-nucleotide polymorphism haplotypes of this gene have been shown to increase the risk for idiopathic generalized epilepsy. Alternatively spliced transcript variants encoding different isoforms found for this gene.
Catalog No. NC3787982 Supplier AURORA BIOLABS LLC Supplier No. 180002100
Mitochondrial NAD-dependent malic enzyme (ME2) is a homotetrameric protein that catalyzes the oxidative decarboxylation of malate to pyruvate. It had previously been weakly linked to a syndrome known as Friedreich ataxia that has since been shown to be the result of mutation in a completely different gene. Certain single-nucleotide polymorphism haplotypes of this gene have been shown to increase the risk for idiopathic generalized epilepsy. Alternatively spliced transcript variants encoding different isoforms found for this gene.